Merck
  • Genome sequence of Hydrogenophaga sp. strain PBC, a 4-aminobenzenesulfonate-degrading bacterium.
Genome sequence of Hydrogenophaga sp. strain PBC, a 4-aminobenzenesulfonate-degrading bacterium.
Journal of bacteriology (2012-08-14)
Han Ming Gan, Teong Han Chew, Yea-Ling Tay, Siew Fen Lye, Adibah Yahya
ABSTRACT
Hydrogenophaga sp. strain PBC is an effective degrader of 4-aminobenzenesulfonate isolated from textile wastewater. Here we present the assembly and annotation of its genome, which may provide further insights into its metabolic potential. This is the first announcement of the draft genome sequence of a strain from the genus Hydrogenophaga.
Sigma-Aldrich
Sulfanilic acid, JIS special grade, 99.0-100.5%
Sigma-Aldrich
Sulfanilic acid, ACS reagent, 99%
Sigma-Aldrich
Sulfanilic acid, puriss. p.a., ≥99.0% (T)
Millipore
Nitrate Reagent B, suitable for microbiology