Merck
  • DNA-binding, spectroscopic and antimicrobial studies of palladium(II) complexes containing 2,2'-bipyridine and 1-phenylpiperazine.
DNA-binding, spectroscopic and antimicrobial studies of palladium(II) complexes containing 2,2'-bipyridine and 1-phenylpiperazine.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-08-07)
Azza A Shoukry, Mervat S Mohamed
ABSTRACT
With the purpose of evaluating the ability of Pd(II) complex to interact with DNA molecule as the main biological target, two new complexes [Pd(bpy)(OH(2))(2)] (1) and [Pd(Phenpip)(OH(2))(2)] (2), where (bpy=2,2'-bipyridine; Phenpip=1-phenylpiperazine), have been synthesized and the binding properties of these complexes with CT-DNA were investigated. The intrinsic binding constants (K(b)) calculated from UV-Vis absorption studies were 3.78×10(3) M(-1) and 4.14×10(3)M(-1) for complexes 1 and 2 respectively. Thermal denaturation has been systematically studied by spectrophotometric method and the calculated ΔT(m) was nearly 5 °C for each complex. All the results suggest an electrostatic and/or groove binding mode for the interaction between the complexes and CT-DNA. The redox behavior of the two complexes in the absence and in the presence of calf thymus DNA has been investigated by cyclic voltammetry. The cyclic voltammogram exhibits one quasi-reversible redox wave. The change in E(1/2), ΔE(p) and I(pc)/I(pa) supports that the two complexes exhibit strong binding to calf thymus DNA. Further insight into the binding of complexes with CT-DNA has been made by gel electrophoresis, where the binding of complexes is confirmed through decreasing the intensity of DNA bands. The two complexes have been screened for their antimicrobial activities using the disc diffusion method against some selected Gram-positive and Gram-negative bacteria. The activity data showed that both complexes were more active against Gram-negative than Gram-positive bacteria. It may be concluded that the antimicrobial activity of the compounds is related to cell wall structure of bacteria.
Sigma-Aldrich
1-Phenylpiperazine, ≥97.0%
Pricing and availability is not currently available.