Merck
Resistance to fracture of roots filled with different sealers.
Dental materials journal (2012-08-07)
Burak Sağsen, Yakup Ustün, Kanşad Pala, Sezer Demırbuğa
ABSTRACT
The aim of this study was to compare the fracture resistance of roots filled with gutta percha (GP) and different root canal sealers.Fifty-five human maxillary central incisors were selected and randomly divided into three experimental groups (Groups 1-3) and two control groups (Groups 4 and 5). They were Group 1-15 root canals filled with an epoxy resin-based sealer (AH Plus) and GP, Group 2 -15 root canals filled with a calcium silicate-based sealer (iRoot SP) and GP, Group 3: 15 root canals filled with another calcium silicate-based sealer (MTA Fillapex) and GP, Group 4: five roots were instrumented but not filled, and Group 5: five roots were neither instrumented nor filled. Compressive loading was carried out using a universal testing machine until fracture occurred. Force applied at time of fracture was recorded as fracture strength of specimen in Newtons. There were no significant differences in fracture strength among the three experimental groups (p>0.05), whose results were significantly superior to that of Group 4 (p<0.05). In conclusion, all the root canal sealers used in the present study increased the fracture resistance of instrumented root canals.
Sigma-Aldrich
Calcium silicate, −200 mesh, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium silicate, purum, 12-22% Ca (as CaO) basis, ≥87% SiO2 basis
Pricing and availability is not currently available.