Merck
  • Development of a vitrification-based cryopreservation protocol for the storage of saltcedar (Tamarix boveana Bunge).
Development of a vitrification-based cryopreservation protocol for the storage of saltcedar (Tamarix boveana Bunge).
Cryo letters (2012-07-25)
M Cano-Castillo, J L Casas
ABSTRACT
We cryopreserved in vitro shoot tips of saltcedar (Tamarix boveana Bunge) using the vitrification technique. The success of the cryopreservation protocol was strongly affected by preculture, loading duration, dehydration duration in plant vitrification solution 2 (PVS2), and medium composition during post-warming regrowth. The highest explant regrowth (50 percent) occurred when the following conditions were employed: preculture in 0.4 M glycerol; treatment with a loading solution (LS) consisting of 2 M glycerol + 0.4 M sucrose in culture medium for 40 min at room temperature; and dehydration in PVS2 at 0 degree C for 45 min before rapid immersion in liquid nitrogen (LN). Rewarming was performed in a water-bath at 40 degree C for 2 min. Explants were then immersed in unloading solution for 10 min before plating on recovery medium supplemented with 0.01 mg per liter thidiazuron (TDZ). TDZ was progressively eliminated from the medium over a period of 6 weeks. Plantlets were transferred to a double-layer medium to enhance rooting. This protocol was successfully applied to three individuals of T. boveana harvested from the wild.
Product Description
Supelco
Thidiazuron, PESTANAL®, analytical standard
Sigma-Aldrich
Thidiazuron, suitable for plant cell culture, BioReagent