Merck
  • Single bout of endurance exercise increases NNMT activity in the liver and MNA concentration in plasma; the role of IL-6.
Single bout of endurance exercise increases NNMT activity in the liver and MNA concentration in plasma; the role of IL-6.
Pharmacological reports : PR (2012-06-05)
Stefan Chłopicki, Marta Kurdziel, Magdalena Sternak, Małgorzata Szafarz, Joanna Szymura-Oleksiak, Karol Kamiński, Jerzy A Żołądź
ABSTRACT
Methylnicotinamide (MNA) displays vasoprotective activity, however, the regulation of the activity of nicotinamide-N-methyltransferase (NNMT), is largely unknown. We analyze a possible involvement of IL-6 in the activation of NNMT-MNA pathway during an endurance exercise. FVB, C57Bl/6J IL6(+/+) and C57Bl/6J IL-6(-/-) mice were subjected to the single bout of endurance exercise consisting of 90 min of swimming. Thereafter, exercise-induced changes in NNMT activity in the liver as well as concomitant changes in the concentration of MNA and its further metabolites in plasma were analyzed. In two strains of mice (FVB and C57Bl/6J IL6(+/+)) 90 min of swimming resulted in approximately 2-3 folds increase in NNMT activity (from 0.14 ± 0.03 to 0.421 ± 0.02 pmol/min/mg, p < 0.05 and from 0.2 ± 0.06 to 0.35 ± 0.07 pmol/min/mg, p < 0.01, respectively) and concomitant increase in the plasma concentration of MNA (from 157 ± 15.06 to 230 ± 16.2 ng/ml, p < 0.01, and from 77.05 ± 14.6 ng/ml to 152.55 ± 58.4 ng/ml; p < 0.01, respectively). However, in C57Bl/6J IL-6(-/-) mice 90 min of swimming did not change liver NNMT activity (from 0.25 ± 0.07 to 0.23 ± 0.06 pmol/min/mg), while MNA concentration in plasma rose approximately two-fold (from 65.3 ± 30.9 ng/ml to 124.8 ± 35.8 ng/ml; p < 0.05). We demonstrated for the first time that NNMT - MNA pathway is activated by a single bout of endurance exercise. Interestingly, exercise-induced activation of NNMT in the liver involves IL-6, while the rise in MNA concentration in plasma was partially IL-6-independent. Taking into the consideration the pharmacological activity of MNA, IL-6-dependent and IL-6-independent activation of NNMT, may contribute to the exercise capacity. The physiological role of NNMT in the exercise warrant further studies.
Product Description
Supelco
1-Methylnicotinamide chloride, analytical standard
Pricing and availability is not currently available.