Merck
  • The 894T allele of endothelial nitric oxide synthase gene is related to left ventricular mass in African Americans with high-normal blood pressure.
The 894T allele of endothelial nitric oxide synthase gene is related to left ventricular mass in African Americans with high-normal blood pressure.
Journal of the National Medical Association (2005-02-17)
Rigobert Lapu-Bula, Alexander Quarshie, Deborah Lyn, Adefisayo Oduwole, Cheryl Pack, Jan Morgan, Sunday Nkemdiche, Priscilla Igho-Pemu, Anekwe Onwuanyi, Rongling Li, Elizabeth Ofili
ABSTRACT
The 894T allele in exon 7 of the endothelial nitric oxide synthase (eNOS) gene has been inconsistently associated with hypertension in different racial groups. Because high-normal blood pressure (BP) confers an increased risk for the development of hypertension and other cardiovascular disorders, including left ventricular hypertrophy (LVH), we tested the hypothesis that the allelic variation (894T) in the eNOS gene would directly correlate with alterations in LV mass (LVM) in individuals with high-normal BP. Genotype distribution of G894T was compared between 20 African Americans (10 females/10 males) with high-normal BP (systolic BP of 130-139 and/or diastolic BP of 85-89 mmHg) and 64 counterparts (37 females/27 males) with normal BP (<130/85 mmHg). Echocardiographic LVM was calculated (Devereux formula) and indexed to body surface area to define the presence of LVH (LVMI >134/110 g/m2 for men/women). For the entire group, the 894T allelic frequencies (15, 48%) and G894T genotype distributions were consistent with the Hardy-Weinberg equilibrium expectations (estimated disequilibrium coefficient = 0.0118, P=0.40). LVMI was significantly higher in homozygous carriers (TT) of the rare 894T allele (n = 3 females/0 males) than in heterozygous GT (n = 13 females/7 males) and individuals bearing the GG (n=34 females/27 males) variant (124 +/- 70 vs. 82 +/- 24 and 82 +/- 19 g/m2, respectively, P < 0.05). The observed relationship between eNOS 894T allele and LVMI was restricted to individuals with high-normal BP (r = 0.94, P = 0.03) but not in those with normal BP (r = 0.39, P =0.64), by analysis of variance (ANOVA) after adjusting for age, gender, body mass index, smoking and systolic BP. These findings, not previously described, provide important preliminary evidence to suggest an increased susceptibility to LVH in African Americans who carry the 894T variant of the eNOS gene and have high-normal blood pressure.