Merck
  • Treatment of herpes labialis: comparison of two OTC drugs and untreated controls.
Treatment of herpes labialis: comparison of two OTC drugs and untreated controls.
Journal of esthetic and restorative dentistry : official publication of the American Academy of Esthetic Dentistry ... [et al.] (2012-04-25)
James P McCarthy, William D Browning, Craig Teerlink, George Veit
ABSTRACT
Rapid resolution of active herpes labialis lesions is of great benefit to the patient not only in terms of controlling pain and disfigurement, but in disruption of needed dental treatment. Using three groups, this retrospective study investigated the time required to complete healing and the loss of discomfort. Based on 180 completed surveys, responses were divided into three groups: One group used Abreva (GlaxoSmithKline, Parsippany, NJ, USA). The second used Viroxyn (Quadex Pharmaceuticals, West Jordan, UT, USA). The third group, the Control group, consisted of untreated lesions. All three groups were asked about past experiences with lesions treated using Abreva and/or Viroxyn, and lesions which were left untreated. In addition, 58 participants who had used Viroxyn only responded. Participants were provided standardized responses from which to choose. For both the time to healing and time to loss of discomfort, participants in both the Abreva and Viroxyn groups experienced significant improvements. Relative to the Abreva, Viroxyn provided significant improvement in both outcomes (all t-tests; all p < 0.001). Relative to the Control group, Viroxyn and Abreva offered an 8.0 and 4.0 day reduction in time to healing, respectively. Loss of discomfort occurred within 3.0 days and 1 hour for Abreva and Viroxyn, respectively. Relative to the untreated controls, both Abreva and Viroxyn offered a significant reduction in both the time to healing and time to loss of discomfort. Furthermore, Viroxyn offered a significant reduction relative to Abreva. .
Sigma-Aldrich
1-Docosanol, 98%