Merck
  • Kinetic effects of tartaric acid on the growth of chiral J-aggregates of tetrakis(4-sulfonatophenyl)porphyrin.
Kinetic effects of tartaric acid on the growth of chiral J-aggregates of tetrakis(4-sulfonatophenyl)porphyrin.
Chemical communications (Cambridge, England) (2012-04-14)
Maria Angela Castriciano, Andrea Romeo, Roberto Zagami, Norberto Micali, Luigi Monsù Scolaro
ABSTRACT
The kinetics of growth for chiral J-aggregates of H(4)TPPS(4) porphyrin have been investigated under different experimental conditions in the presence of tartaric acid. The observed rate constants and the anisotropy factor g show a defined dependence on the enantiomer used as a chiral templating agent.
Product Description
Supelco
4,4′,4″,4″′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzenesulfonic acid), for spectrophotometric det. of transition metals, ≥95.0%