Merck
  • The role of molecular regulation and targeting in regulating calcium/calmodulin stimulated protein kinases.
The role of molecular regulation and targeting in regulating calcium/calmodulin stimulated protein kinases.
Advances in experimental medicine and biology (2012-03-29)
Kathryn A Skelding, John A P Rostas
ABSTRACT
Calcium/calmodulin-stimulated protein kinases can be classified as one of two types - restricted or multifunctional. This family of kinases contains several structural similarities: all possess a calmodulin binding motif and an autoinhibitory region. In addition, all of the calcium/calmodulin-stimulated protein kinases examined in this chapter are regulated by phosphorylation, which either activates or inhibits their kinase activity. However, as the multifunctional calcium/calmodulin-stimulated protein kinases are ubiquitously expressed, yet regulate a broad range of cellular functions, additional levels of regulation that control these cell-specific functions must exist. These additional layers of control include gene expression, signaling pathways, and expression of binding proteins and molecular targeting. All of the multifunctional calcium/calmodulin-stimulated protein kinases examined in this chapter appear to be regulated by these additional layers of control, however, this does not appear to be the case for the restricted kinases.
Sigma-Aldrich
Phosphorylase Kinase from rabbit muscle, lyophilized powder, ≥60 units/mg protein
Pricing and availability is not currently available.