Merck
  • Rapid detection, differentiation and typing of methicillin-resistant Staphylococcus aureus harbouring either mecA or the new mecA homologue mecA(LGA251).
Rapid detection, differentiation and typing of methicillin-resistant Staphylococcus aureus harbouring either mecA or the new mecA homologue mecA(LGA251).
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases (2012-03-21)
M Stegger, P S Andersen, A Kearns, B Pichon, M A Holmes, G Edwards, F Laurent, C Teale, R Skov, A R Larsen
ABSTRACT
The recent finding of a new mecA homologue, mecA(LGA251) , with only 70% nucleotide homology to the conventional mecA gene has brought the routine testing for mecA as a confirmatory test for methicillin-resistant Staphylococcus aureus (MRSA) into question. A multiplex PCR was designed to differentiate mecA(LGA251) from the known mecA together with detection of lukF-PV and the spa gene fragments, enabling direct spa typing by sequencing of the PCR amplicons. The PCR analysis and subsequent spa typing were validated on a large collection (n=185) of contemporary MRSA and methicillin-sensitive S. aureus isolates, including 127 isolates carrying mecA(LGA251) . The mecA(LGA251) gene was situated in staphylococcal cassette chromosome mec type XI elements, and sequence variation within a 631-bp fragment of mecA(LGA251) in 79 isolates indicated a very conserved gene sequence. Following a successful validation, the multiplex PCR strategy was implemented in the routine testing of MRSA for national surveillance. Over a 2-month period, among 203 samples tested, 12 new MRSA cases caused by isolates carrying mecA(LGA251) were identified, emphasizing the clinical importance of testing for these new MRSA isolates.
Product Description
Cefoxitin sodium, European Pharmacopoeia (EP) Reference Standard
Supelco
Cefoxitin sodium salt