Merck
Sotalol permeability in cultured-cell, rat intestine, and PAMPA system.
Pharmaceutical research (2012-02-22)
Wei Liu, Hideaki Okochi, Leslie Z Benet, Suo-Di Zhai
ABSTRACT
To clarify sotalol's classification in the BCS versus BDDCS systems through cellular, rat everted sac and PAMPA permeability studies. Studies were carried out in Madin Darby canine kidney (MDCK) and MDR1-transfected MDCK (MDCK-MDR1) cell lines, rat everted gut sacs and the Parallel Artificial Membrane Permeability Assay (PAMPA) system. Three-hour transport studies were conducted in MDCK cell lines (with apical pH changes) and MDCK-MDR1 cells (with and without the P-glycoprotein inhibitor GG918); male Sprague-Dawley rats (300~350 g) were used to prepare everted sacs. In the PAMPA studies, drug solutions at different pH's were dosed in each well and incubated for 5 h. Samples were measured by LC-MS/MS, or liquid scintillation counting and apparent permeability (P(app)) was calculated. Sotalol showed low permeability in all of the cultured-cell lines, everted sacs and PAMPA systems. It might be a border line P-glycoprotein substrate. The PAMPA study showed that sotalol's permeability increased with a higher apical pH, while much less change was found in MDCK cells. The low permeability rate for sotalol correlates with its Class 3 BDDCS assignment and lack of in vivo metabolism.
Sigma-Aldrich
(±)-Sotalol hydrochloride, ≥98% (TLC), powder
Pricing and availability is not currently available.