Merck
  • Enhancement of biodiesel synthesis from soybean oil by potassium fluoride modification of a calcium magnesium oxides catalyst.
Enhancement of biodiesel synthesis from soybean oil by potassium fluoride modification of a calcium magnesium oxides catalyst.
Bioresource technology (2011-12-16)
Mingming Fan, Pingbo Zhang, Qinke Ma
ABSTRACT
Transesterification of soybean oil with methanol was carried out in the presence of CaO-MgO and KF-modified CaO-MgO catalysts at atmospheric pressure. While the methyl ester yield for the CaO-MgO catalyst with a ratio of 8:2 (CaO:MgO) was 63.6%, it was 97.9% for the KF-modified catalyst at a 2% catalyst to the reactants (methanol/oil mixture) weight ratio, a temperature of 65 °C, a methanol-soybean oil ratio of 9:1 and a reaction time of 2.5 h. The KF/CaO-MgO catalyst still yielded 86.7% after four successive uses. The catalytic performance of the KF/CaO-MgO catalyst was attributed to the formation of active KCaF(3) and K(2)MgF(4) centers.
Sigma-Aldrich
Potassium fluoride, SAJ first grade, ≥95.0%
Sigma-Aldrich
Potassium fluoride, anhydrous, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Potassium fluoride, ≥99.97% trace metals basis
Sigma-Aldrich
Potassium fluoride, BioUltra, ≥99.5% (F)