Merck
  • Geranylgeranylacetone, heat shock protein 90/AMP-activated protein kinase/endothelial nitric oxide synthase/nitric oxide pathway, and endothelial function in humans.
Geranylgeranylacetone, heat shock protein 90/AMP-activated protein kinase/endothelial nitric oxide synthase/nitric oxide pathway, and endothelial function in humans.
Arteriosclerosis, thrombosis, and vascular biology (2011-10-15)
Noritaka Fujimura, Daisuke Jitsuiki, Tatsuya Maruhashi, Shinsuke Mikami, Yumiko Iwamoto, Masato Kajikawa, Kazuaki Chayama, Yasuki Kihara, Kensuke Noma, Chikara Goto, Yukihito Higashi
ABSTRACT
Geranylgeranylacetone (GGA) induces expression of heat shock protein 90 (Hsp90), an adaptor molecule for assembly of endothelial nitric oxide synthase (eNOS) phosphorylation complex. The purpose of this study was to determine whether GGA enhances Hsp90 expression and augments endothelium-dependent vasodilation via upregulation of eNOS in humans. We evaluated the effects of GGA on human umbilical vein endothelial cells (HUVECs) and on forearm blood flow (FBF) responses to acetylcholine and sodium nitroprusside in 40 healthy young men. Hsp90, eNOS, AMP-activated protein kinase (AMPK), and Akt expression in HUVECs and peripheral blood mononuclear cells was detected by Western blot analysis. GGA increased Hsp90 expression and phosphorylation of eNOS and AMPK but not Akt in HUVECs and increased Hsp90 expression in peripheral blood mononuclear cells. Oral administration of GGA (600 mg) augmented the FBF response to acetylcholine. Infusion of N(G)-monomethyl-l-arginine, an NO synthase inhibitor, completely abolished GGA-induced augmentation of the FBF response to acetylcholine. GGA also augmented the acetylcholine-stimulated NO release in smokers. These findings suggest that GGA-induced activation of Hsp90/AMPK significantly increased NO-mediated vasodilation in healthy subjects, as well as in smokers. The use of GGA may be a new therapeutic approach for improving endothelial dysfunction.
Sigma-Aldrich
Geranylgeranylacetone
Pricing and availability is not currently available.
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Pricing and availability is not currently available.