Merck
  • Approaches to efficient molecular catalyst systems for photochemical H2 production using [FeFe]-hydrogenase active site mimics.
Approaches to efficient molecular catalyst systems for photochemical H2 production using [FeFe]-hydrogenase active site mimics.
Dalton transactions (Cambridge, England : 2003) (2011-10-11)
Mei Wang, Lin Chen, Xueqiang Li, Licheng Sun
ABSTRACT
The research on structural and functional biomimics of the active site of [FeFe]-hydrogenases is in an attempt to elucidate the mechanisms of H(2)-evolution and uptake at the [FeFe]-hydrogenase active site, and to learn from Nature how to create highly efficient H(2)-production catalyst systems. Undoubtedly, it is a challenging, arduous, and long-term work. In this perspective, the progresses in approaches to photochemical H(2) production using mimics of the [FeFe]-hydrogenase active site as catalysts in the last three years are reviewed, with emphasis on adjustment of the redox potentials and hydrophilicity of the [FeFe]-hydrogenase active site mimics to make them efficient catalysts for H(2) production. With gradually increasing understanding of the chemistry of the [FeFe]-hydrogenases and their mimics, more bio-inspired proton reduction catalysts with significantly improved efficiency of H(2) production will be realized in the future.
Sigma-Aldrich
Iron(II) sulfide, SAJ first grade, ≥50.0%
Sigma-Aldrich
Iron(II) sulfide, technical grade
Sigma-Aldrich
Iron(II) sulfide, −100 mesh, 99.9% trace metals basis