Merck
  • Synthesis and characterization of lawsone-lntercalated Zn-Al-layered double hydroxides.
Synthesis and characterization of lawsone-lntercalated Zn-Al-layered double hydroxides.
Journal of biomedical nanotechnology (2011-08-13)
Yamin Yasin, Nur Mushirah Ismail, Mohd Zobir Hussein, Norhaniza Aminudin
ABSTRACT
A drug-inorganic nanostructured material involving pharmaceutically active compound lawsone intercalated Zn-Al layered double hydroxides (Law-LDHs) with Zn/AI = 4 has been assembled by co-precipitation and ion exchange methods. Powder X-ray diffraction (XRD) and Fourier transform infrared spectra (FTIR) analysis indicate a successful intercalation of lawsone between the layers of layered double hydroxides. It suggests that layered double hydroxides may have application as the basis of a drug delivery system.
Sigma-Aldrich
2-Hydroxy-1,4-naphthoquinone, 97%
Pricing and availability is not currently available.