Merck
  • An alkaline β-glucosidase isolated from an olive brine strain of Wickerhamomyces anomalus.
An alkaline β-glucosidase isolated from an olive brine strain of Wickerhamomyces anomalus.
FEMS yeast research (2011-05-18)
Cristina Restuccia, Serena Muccilli, Rosa Palmeri, Cinzia L Randazzo, Cinzia Caggia, Giovanni Spagna
ABSTRACT
An efficient β-glucosidase (βG)-producing strain, Wickerhamomyces anomalus BS81, was isolated from naturally fermented olive brine and identified based on PCR/restriction fragment length polymorphism of the rDNA internal transcribed spacer and sequence analysis of the D1/D2 region of the 26S rRNA gene. The hydrolytic activity of the βG had an optimum pH of 8.5 and an optimum temperature of 35 °C. The enzyme had high substrate specificity and high catalytic efficiency (K(m) 0.99 mM, V(max) 14 U g(-1) of cells) for p-nitrophenyl-β-d-glucopyranoside. The enzyme was activated by increasing concentrations of NaCl, with maximum activity at 150 g L(-1) NaCl. Although βGs have been purified and characterized from several other sources, the W. anomalusβG is unique among βGs because its relative maximum activity occurs at alkaline pH and 35 °C. Moreover, the yeast strain has esterase activity that acts synergistically with βG to degrade oleuropein to debitter table olives and olive oil.
Sigma-Aldrich
4-Nitrophenyl β-D-glucopyranoside, ≥98% (TLC)