Merck
  • Humic acid coated Fe3O4 magnetic nanoparticles as highly efficient Fenton-like catalyst for complete mineralization of sulfathiazole.
Humic acid coated Fe3O4 magnetic nanoparticles as highly efficient Fenton-like catalyst for complete mineralization of sulfathiazole.
Journal of hazardous materials (2011-04-26)
Hongyun Niu, Di Zhang, Shengxiao Zhang, Xiaole Zhang, Zhaofu Meng, Yaqi Cai
ABSTRACT
Humic acid coated Fe(3)O(4) magnetic nanoparticles (Fe(3)O(4)/HA) were prepared for the removal of sulfathiazole from aqueous media. Fe(3)O(4)/HA exhibited high activity to produce hydroxyl (OH) radicals through catalytic decomposition of H(2)O(2). The degradation of sulfathiazole was strongly temperature-dependent and favored in acidic solution. The catalytic rate was increased with Fe(3)O(4)/HA dosage and H(2)O(2) concentration. When 3 g L(-1) of Fe(3)O(4)/HA and 0.39 M of H(2)O(2) were introduced to the aqueous solution, most sulfathiazole was degraded within 1h, and >90% of total organic carbon (TOC) were removed in the reaction period (6h). The major final products were identified as environmentally friendly ions or inorganic molecules (SO(4)(2-), CO(2), and N(2)). The corresponding degradation rate (k) of sulfathiazole and TOC was 0.034 and 0.0048 min(-1), respectively. However, when 3 g L(-1) of bare Fe(3)O(4) were used as catalyst, only 54% of TOC was eliminated, and SO(4)(2-) was not detected within 6h. The corresponding degradation rate for sulfathiazole and TOC was 0.01 and 0.0016 min(-1), respectively. The high catalytic ability of Fe(3)O(4)/HA may be caused by the electron transfer among the complexed Fe(II)-HA or Fe(III)-HA, leading to rapid regeneration of Fe(II) species and production of OH radicals.
Product Description
Supelco
Sulfathiazole
Pricing and availability is not currently available.
Supelco
Sulfathiazole, VETRANAL®, analytical standard
Pricing and availability is not currently available.