Merck
  • A highly selective colorimetric and ratiometric two-photon fluorescent probe for fluoride ion detection.
A highly selective colorimetric and ratiometric two-photon fluorescent probe for fluoride ion detection.
Organic letters (2011-02-09)
Jun Feng Zhang, Chang Su Lim, Sankarprasad Bhuniya, Bong Rae Cho, Jong Seung Kim
ABSTRACT
A naphthalimide-based highly selective colorimetric and ratiometric fluorescent probe for the fluoride ion displayed both one- and two-photon ratiometric changes. Upon reaction with the F(-) (TBA(+) and Na(+) salts) anion in CH(3)CN as well as in aqueous buffer solution, probe 1 shows dramatic color changes from colorless to jade-green and remarkable ratiometric fluorescence enhancements signals. These properties are mechanistically ascribed to a fluoride-triggered Si-O bond cleavage that resulted in a green fluorescent 4-amino-1,8-naphthalimide.
Sigma-Aldrich
4-Amino-1,8-naphthalimide