Merck
  • Vinyl diazophosphonates as precursors to quaternary substituted indolines and cyclopentenes.
Vinyl diazophosphonates as precursors to quaternary substituted indolines and cyclopentenes.
Organic letters (2011-01-18)
Jin Wang, Vyacheslav Boyarskikh, Jon D Rainier
ABSTRACT
Vinyl diazophosphonates can be stereoselectively synthesized and, depending upon their substitution pattern, undergo intramolecular C-H insertion reactions or sulfonium ylide rearrangements when exposed to Rh(2)(OAc)(4).
Sigma-Aldrich
Rhodium(II) acetate dimer, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Rhodium(II) acetate dimer, 99.9% trace metals basis