Merck
IL-4 mediates dicloxacillin-induced liver injury in mice.
Toxicology letters (2010-11-26)
Satonori Higuchi, Masanori Kobayashi, Yukitaka Yoshikawa, Koichi Tsuneyama, Tatsuki Fukami, Miki Nakajima, Tsuyoshi Yokoi
ABSTRACT
Drug-induced liver injury (DILI) is a major problem in drug development and clinical drug therapy. In most cases, the mechanisms are still unknown. It is difficult to predict DILI in humans due to the lack of experimental animal models. Dicloxacillin, penicillinase-sensitive penicillin, rarely causes cholestatic or mixed liver injury, and there is some evidence for immunoallergic idiosyncratic reaction in human. In this study, we investigated the mechanisms of dicloxacillin-induced liver injury. Plasma ALT and total-bilirubin (T-Bil) levels were significantly increased in dicloxacillin-administered (600 mg/kg, i.p.) mice. Dicloxacillin administration induced Th2 (helper T cells)-mediated factors and increased the plasma interleukin (IL)-4 level. Neutralization of IL-4 suppressed the hepatotoxicity of dicloxacillin, and recombinant mouse IL-4 administration (0.5 or 2.0 μg/mouse, i.p.) exacerbated it. Chemoattractant receptor-homologous molecule expressed on Th2 cells (CRTh2) is a cognate receptor for prostaglandin (PG) D(2), and is suggested to be involved in Th2-dependent allergic inflammation. We investigated the effect of 13,14-Dihydro-15-keto-PGD(2) (DK-PGD(2); 10 μg/mouse, i.p.) administration on dicloxacillin-induced liver injury. DK-PGD(2)/dicloxacillin coadministration resulted in a significant increase of alanine aminotransferases and a remarkable increase of macrophage inflammatory protein 2 expression. In conclusion, to the best of our knowledge, this is the first report to demonstrate that dicloxacillin-induced liver injury is mediated by a Th2-type immune reaction and exacerbated by DK-PGD(2).
Sigma-Aldrich
Dicloxacillin sodium salt monohydrate
Pricing and availability is not currently available.