Merck
  • Application of sugaring-out extraction for the determination of sulfonamides in honey by high-performance liquid chromatography with fluorescence detection.
Application of sugaring-out extraction for the determination of sulfonamides in honey by high-performance liquid chromatography with fluorescence detection.
Journal of chromatography. A (2010-11-04)
Wen-Hsien Tsai, Hung-Yi Chuang, Ho-Hsien Chen, Yuh-Wern Wu, Shou-Hsun Cheng, Tzou-Chi Huang
ABSTRACT
A simple sugaring-out assisted liquid-liquid extraction method combined with high-performance liquid-chromatography with fluorescence detection (HPLC-FL) was developed for the extraction and determination of sulfonamides in honey. Sample preparation consisted of acid hydrolysis to release sugar-bound sulfonamides. After derivatization with fluorescamine, the derivatives were partitioned into the organic layer under the honey (sugar)/water/acetonitrile system. The clear organic extract obtained by centrifugation could be injected into the HPLC system either directly or after dilution. Linearity was obtained with the coefficient of determination (R(2)) higher than 0.998 from 2 to 200 ng/mL. Under the optimal conditions, recoveries were determined for honey fortified at three levels (5, 20, and 100 ng/g) were 80.9-99.6% with coefficients of variation of 0.3-4.4%. Limits of detection for the sulfonamides studied were found to range from 0.6 to 0.9 ng/g.
Sigma-Aldrich
Fluorescamine, ≥98.0%
Sigma-Aldrich
Fluorescamine, ≥98% (TLC), powder, used for detection of primary amines