Merck
  • Hydrothermal synthesis of TiO2 hollow microspheres for the photocatalytic degradation of 4-chloronitrobenzene.
Hydrothermal synthesis of TiO2 hollow microspheres for the photocatalytic degradation of 4-chloronitrobenzene.
Journal of hazardous materials (2010-09-22)
Miaomiao Ye, Zhonglin Chen, Wenshou Wang, Jimin Shen, Jun Ma
ABSTRACT
TiO(2) hollow microspheres were synthesized by a simple hydrothermal method followed by calcination at different temperatures ranging from 400 to 800°C. The prepared samples were characterized by XRD, SEM, TEM, SAED, HRTEM, N(2) adsorption, and UV-vis spectroscopy. The photocatalytic activities of the hollow microspheres were evaluated by photocatalytic decomposition of 4-chloronitrobenzene (4-CNB). Results showed that the TiO(2) hollow microspheres, which had an average external diameter of 1.75 μm, were composed of numerous TiO(2) nanoparticles. Photocatalysis experiments indicated that the TiO(2) hollow microspheres calcined at 500°C exhibited the highest photocatalytic activity, which was nearly 2 and 1.5 times higher than that of the uncalcined sample and Degussa P25, respectively. The catalyst crystallinity, catalyst dosage and 4-CNB concentration were found to have a significant impact on the degradation efficiency whereas solution pH has relatively less effect. The removal of total organic carbon (TOC) and formation of chloride, nitrate (V) anions were monitored to follow the mineralization process of 4-CNB. In addition, it was demonstrated that these TiO(2) hollow microspheres could be recycled easily without decreasing their photocatalytic activities.
Sigma-Aldrich
1-Chloro-4-nitrobenzene, 99%
Pricing and availability is not currently available.