Merck
  • Cu(OAc)2/MCM-41: an efficient and solid acid catalyst for synthesis of 2-arylbenzothiazoles under ultrasound irradiation.
Cu(OAc)2/MCM-41: an efficient and solid acid catalyst for synthesis of 2-arylbenzothiazoles under ultrasound irradiation.
Ultrasonics sonochemistry (2010-09-02)
Sodeh Sadjadi, Hamid Sepehrian
ABSTRACT
A new, convenient method for the syntheses of 2-arylbenzothiazoles is described in the presence of catalytic amounts of Cu(OAc)(2)/MCM-41 under ultrasonic irradiation. Short reaction times, easy and quick isolation of the products, reusability of the catalyst and excellent yields are the main advantages of this procedure.
Sigma-Aldrich
Copper(II) acetate, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Copper(II) acetate, powder, 99.99% trace metals basis