Merck
  • Determination of octylphenol and nonylphenol in aqueous sample using simultaneous derivatization and dispersive liquid-liquid microextraction followed by gas chromatography-mass spectrometry.
Determination of octylphenol and nonylphenol in aqueous sample using simultaneous derivatization and dispersive liquid-liquid microextraction followed by gas chromatography-mass spectrometry.
Journal of chromatography. A (2010-07-10)
Shusheng Luo, Ling Fang, Xiaowei Wang, Hongtao Liu, Gangfeng Ouyang, Chongyu Lan, Tiangang Luan
ABSTRACT
A simple and fast sample preparation method for the determination of nonylphenol (NP) and octylphenol (OP) in aqueous samples by simultaneous derivatization and dispersive liquid-liquid microextraction (DLLME) was investigated using gas chromatography-mass spectrometry (GC/MS). In this method, a combined dispersant/derivatization catalyst (methanol/pyridine mixture) was firstly added to an aqueous sample, following which a derivatization reagent/extraction solvent (methyl chloroformate/chloroform) was rapidly injected to combine in situ derivatization and extraction in a single step. After centrifuging, the sedimented phase containing the analytes was injected into the GC port by autosampler for analysis. Several parameters, such as extraction solvent, dispersant solvent, amount of derivatization reagent, derivatization and extraction time, pH, and ionic strength were optimized to obtain higher sensitivity for the detection of NP and OP. Under the optimized conditions, good linearity was observed in the range of 0.1-1000 μg L⁻¹ and 0.01-100 μg L⁻¹ with the limits of detection (LOD) of 0.03 μg L⁻¹ and 0.002 μg L⁻¹ for NP and OP, respectively. Water samples collected from the Pearl River were analyzed with the proposed method, the concentrations of NP and OP were found to be 2.40 ± 0.16 μg L⁻¹ and 0.037 ± 0.001 μg L⁻¹, respectively. The relative recoveries of the water samples spiked with different concentrations of NP and OP were in the range of 88.3-106.7%. Compared with SPME and SPE, the proposed method can be successfully applied to the rapid and convenient determination of NP and OP in aqueous samples.