Merck
  • Effect of monomethylethanolamine, dimethylethanolamine, gangliosides, isoproterenol, and 2-hydroxyethylhydrazine on the conversion of ethanolamine to methylated products by cultured chick brain neurons.
Effect of monomethylethanolamine, dimethylethanolamine, gangliosides, isoproterenol, and 2-hydroxyethylhydrazine on the conversion of ethanolamine to methylated products by cultured chick brain neurons.
Journal of neurochemistry (1991-06-01)
C Andriamampandry, L Freysz, J N Kanfer, H Dreyfus, R Massarelli
ABSTRACT
The sequential methylation of ethanolamine and its phosphorylated derivatives has been studied with chick neurons in culture in the presence of several pharmacological agents. Incubation with [3H]ethanolamine in the presence of monomethylethanolamine and dimethylethanolamine indicated that in these neurons the preferential conversion to choline-containing compounds is via the methylation of phosphorylethanolamine. The possibility that there are two separate enzymes, i.e., one responsible for the methylation of water-soluble ethanolamine-containing compounds and another for the ethanolamine phospholipids, was examined with agents believed to influence these conversions. Incubation of neurons in the presence of a mixture of exogenous gangliosides at 10(-8) M and 10(-5) M concentrations showed that these neuritogenic compounds stimulate the methylation of phosphatidylethanolamine and decrease that of phosphorylethanolamine. The inhibitor of phosphatidylethanolamine methyltransferase (EC 2.1.1.17), 2-hydroxyethylhydrazine, decreased the conversion of phosphatidylethanolamine to phosphatidylcholine and increased that of phosphorylethanolamine to phosphorylcholine. The possible effects of adrenergic stimulation were studied by the incubation of neurons with isoproterenol at 10(-6) M and 10(-5) M concentrations. There was a reduction of phosphorylethanolamine methylation and a stimulation of that of phosphatidylethanolamine, and these effects were counteracted by the presence of 5 x 10(-5) M propranolol.
Sigma-Aldrich
2-Hydroxyethylhydrazine, ≥95% (GC)