Merck
  • Determination of triazolam and alpha-hydroxytriazolam in guinea pig hair after a single dose.
Determination of triazolam and alpha-hydroxytriazolam in guinea pig hair after a single dose.
Journal of analytical toxicology (2010-03-13)
Qiran Sun, Ping Xiang, Baohua Shen, Hui Yan, Min Shen
ABSTRACT
A sensitive liquid chromatography-tandem mass spectrometry method is presented for determination of triazolam and alpha-hydroxytriazolam in guinea pig hair after a single dose of triazolam. Eighteen guinea pigs were divided into three dosage groups (10, 100, and 500 microg/kg) and administrated a single dose of triazolam intragastrically. Before administration, drug-free hair was shaved from their back. Newly grown hair in shaved area was collected every seven days after administration. About 20 mg of decontaminated hair was cut into small segments and incubated in 2 mL of phosphate buffer (pH 8.4) at 45 degrees C overnight. Triazolam-d(4) and alpha-hydroxytriazolam-d(4) were used as internal standards, and liquid-liquid extraction was performed with 3 mL of ethyl ether. The sample was separated on an Allure propyl PFP column with a mobile phase of acetonitrile/20 mM ammonium acetate (7:3, v/v). Detection was implemented with multiple reaction monitoring mode by an API4000 triple-quadrupole tandem mass spectrometer. Limits of detection for triazolam and alpha-hydroxytriazolam were 1 and 5 pg/mg, respectively. Triazolam and alpha-hydroxytriazolam could only be detected in the first week, and 100 microg/kg was the minimal dosage detectable in guinea pig hair. The concentration of triazolam in hair was related with administration dosage and hair color. alpha-Hydroxytriazolam has a higher concentration than triazolam in guinea pig hair.
Product Description
Supelco
α-Hydroxytriazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Sigma-Aldrich
α-Hydroxytriazolam
Pricing and availability is not currently available.