Merck
  • Penetrating cation/fatty acid anion pair as a mitochondria-targeted protonophore.
Penetrating cation/fatty acid anion pair as a mitochondria-targeted protonophore.
Proceedings of the National Academy of Sciences of the United States of America (2010-01-19)
Fedor F Severin, Inna I Severina, Yury N Antonenko, Tatiana I Rokitskaya, Dmitry A Cherepanov, Elena N Mokhova, Mikhail Yu Vyssokikh, Antonina V Pustovidko, Olga V Markova, Lev S Yaguzhinsky, Galina A Korshunova, Nataliya V Sumbatyan, Maxim V Skulachev, Vladimir P Skulachev
ABSTRACT
A unique phenomenon of mitochondria-targeted protonophores is described. It consists in a transmembrane H(+)-conducting fatty acid cycling mediated by penetrating cations such as 10-(6'-plastoquinonyl)decyltriphenylphosphonium (SkQ1) or dodecyltriphenylphosphonium (C(12)TPP). The phenomenon has been modeled by molecular dynamics and directly proved by experiments on bilayer planar phospholipid membrane, liposomes, isolated mitochondria, and yeast cells. In bilayer planar phospholipid membrane, the concerted action of penetrating cations and fatty acids is found to result in conversion of a pH gradient (DeltapH) to a membrane potential (Deltapsi) of the Nernstian value (about 60 mV Deltapsi at DeltapH = 1). A hydrophobic cation with localized charge (cetyltrimethylammonium) failed to substitute for hydrophobic cations with delocalized charge. In isolated mitochondria, SkQ1 and C(12)TPP, but not cetyltrimethylammonium, potentiated fatty acid-induced (i) uncoupling of respiration and phosphorylation, and (ii) inhibition of H(2)O(2) formation. In intact yeast cells, C(12)TPP stimulated respiration regardless of the extracellular pH value, whereas a nontargeted protonophorous uncoupler (trifluoromethoxycarbonylcyanide phenylhydrazone) stimulated respiration at pH 5 but not at pH 3. Hydrophobic penetrating cations might be promising to treat obesity, senescence, and some kinds of cancer that require mitochondrial hyperpolarization.
Sigma-Aldrich
Tetraphenylphosphonium chloride, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Tetraphenylphosphonium bromide, 97%
Pricing and availability is not currently available.