Merck
  • Phosphorescent sensing of carbon dioxide based on secondary inner-filter quenching.
Phosphorescent sensing of carbon dioxide based on secondary inner-filter quenching.
Analytica chimica acta (2009-11-21)
I M Pérez de Vargas-Sansalvador, M A Carvajal, O M Roldán-Muñoz, J Banqueri, M D Fernández-Ramos, L F Capitán-Vallvey
ABSTRACT
A study of different strategies to prepare phosphorescence-based sensors for gaseous CO(2) determination has been performed. It includes the characterization of different configurations tested, a discussion of the results obtained and possibilities for the future. The optical sensor for gaseous CO(2) is based on changes in the phosphorescence intensity of the platinum octaethylporphyrin (PtOEP) complex trapped both on oxygen-insensitive poly(vinylidene chloride-co-vinyl chloride) (PVCD) membranes and PVCD microparticles, due to the displacement of the alpha-naphtholphthalein acid-base equilibrium with CO(2) concentration. A secondary inner-filter mechanism was tested for the sensor and a full range linearized calibration was obtained by plotting (I(100)-I(0))/(I-I(0)) versus the inverse of the CO(2) concentration, where I(0) and I(100) are the detected luminescence intensities from a membrane exposed to 100% nitrogen and 100% CO(2), respectively, and I at a defined CO(2) concentration. The different configurations tested included the use of membranes containing luminophore and pH-sensitive dye placed on two opposite sides of a transparent support to prevent the observed degradation of the PtOEP complex in the presence of the tetraoctylammonium hydroxide (TOAOH) phase transfer agent, which produced better results regarding stability and sensitivity. The CO(2) gas sensor based on PtOEP homogeneous membranes presented better properties in terms of response time and sensitivity than that based on PtOEP microparticles. With a detection limit of 0.02%, the response time (10-90% maximum signal) is 9 s and the recovery time (90-10%) is 115 s. The lifetime of the membranes for CO(2) sensing preserved in a 94% RH atmosphere and dark conditions is longer than at least 4 months.
Sigma-Aldrich
α-Naphtholphthalein, practical grade
Pricing and availability is not currently available.