Merck
  • Deuterium isotope effect on the toxicokinetics of monomethylamine in the rat.
Deuterium isotope effect on the toxicokinetics of monomethylamine in the rat.
Drug metabolism and disposition: the biological fate of chemicals (1990-07-01)
A J Streeter, R W Nims, P R Sheffels, J A Hrabie, L Ohannesian, Y H Heur, B A Mico, L K Keefer
ABSTRACT
The single-dose toxicokinetics of monomethylamine has been characterized in the rat by HPLC assay of serial blood samples. Biphasic first-order elimination was observed following an iv bolus dose of 19 mumol/kg with a terminal half-life of 19.1 +/- 1.3 min (mean +/- SE, N = 4). The apparent steady state volume of distribution, systemic blood clearance, and renal blood clearance were 1.21 +/- 0.09 liter/kg, 53.4 +/- 3.5 ml/min/kg, and 5.72 +/- 0.53 ml/min/kg, respectively. The administration of an intragastric dose permitted the calculation of the systemic bioavailability of monomethylamine as 69 +/- 3%. Duplicate experiments using the structural analogue with deuterium atoms substituted for hydrogens on the methyl group revealed a much slower elimination of the compound, although ultimately, 5 times as much was excreted unchanged in the urine. Isotope effects calculated as the ratios of terminal half-life, systemic blood clearance, and systemic bioavailability were 1.9, 2.2, and 1.8, respectively.
Sigma-Aldrich
N-Methylurea, 97%
Pricing and availability is not currently available.