Merck
  • Effect of various visible light photoinitiators on the polymerization and color of light-activated resins.
Effect of various visible light photoinitiators on the polymerization and color of light-activated resins.
Dental materials journal (2009-09-02)
Hiroyuki Arikawa, Hideo Takahashi, Takahito Kanie, Seiji Ban
ABSTRACT
The purpose of this study was to investigate effects of various visible light photoinitiators on the polymerization efficiency and color of the light-activated resins. Four photoinitiators, including camphorquinone, phenylpropanedione, monoacrylphosphine oxide (TPO), and bisacrylphosphine oxide (Ir819), were used. Each photoinitiator was dissolved in a Bis-GMA and TEGDMA monomer mixture. Materials were polymerized using dental quartz-tungsten halogen lamp (QTH), plasma-ark lamp and blue LED light-curing units, and a custom-made violet LED light unit. The degree of monomer conversion and CIE L*a*b* color values of the resins were measured using a FTIR and spectral transmittance meter. The degree of monomer conversions of TPO- and Ir819-containing resins polymerized with the violet-LED unit were higher than camphorquinone-containing resin polymerized with the QTH light-curing unit. The lowest color values were observed for the TPO-containing resin. Our results indicate that the TPO photoinitiator and the violet-LED light unit may provide a useful and improved photopolymerization system for dental light-activated resins.
Sigma-Aldrich
1-Phenyl-1,2-propanedione, 98%, FG
Sigma-Aldrich
1-Phenyl-1,2-propanedione, 99%