Merck
  • Alpha-fluorovinyl Weinreb amides and alpha-fluoroenones from a common fluorinated building block.
Alpha-fluorovinyl Weinreb amides and alpha-fluoroenones from a common fluorinated building block.
The Journal of organic chemistry (2009-04-14)
Arun K Ghosh, Shaibal Banerjee, Saikat Sinha, Soon Bang Kang, Barbara Zajc
ABSTRACT
Synthesis and reactivity of N-methoxy-N-methyl-(1,3-benzothiazol-2-ylsulfonyl)fluoroacetamide, a building block for Julia olefination, is reported. This reagent undergoes condensation reactions with aldehydes and cyclic ketones to give alpha-fluorovinyl Weinreb amides. Olefination reactions proceed under mild, DBU-mediated conditions, or in the presence of NaH. DBU-mediated condensations proceed with either E- or Z-selectivity, depending upon reaction conditions, whereas NaH-mediated reactions are > or = 98% Z-stereoselective. Conversion of the Weinreb amide moiety in N-methoxy-N-methyl-(1,3-benzothiazol-2-ylsulfanyl)fluoroacetamide to ketones, followed by oxidation, resulted in another set of olefination reagents, namely (1,3-benzothiazol-2-ylsulfonyl)fluoromethyl phenyl and propyl ketones. In the presence of DBU, these compounds react with aldehydes tested to give alpha-fluoroenones with high Z-selectivity. The use of N-methoxy-N-methyl-(1,3-benzothiazol-2-ylsulfanyl)fluoroacetamide as a common fluorinated intermediate in the synthesis of alpha-fluorovinyl Weinreb amides and alpha-fluoroenones has been demonstrated. Application of the Weinreb amide to alpha-fluoro allyl amine synthesis is also shown.
Sigma-Aldrich
Fluoroacetamide