Merck
A straight path to circular proteins.
The Journal of biological chemistry (2009-04-11)
John M Antos, Maximilian Wei-Lin Popp, Robert Ernst, Guo-Liang Chew, Eric Spooner, Hidde L Ploegh
ABSTRACT
Folding and stability are parameters that control protein behavior. The possibility of conferring additional stability on proteins has implications for their use in vivo and for their structural analysis in the laboratory. Cyclic polypeptides ranging in size from 14 to 78 amino acids occur naturally and often show enhanced resistance toward denaturation and proteolysis when compared with their linear counterparts. Native chemical ligation and intein-based methods allow production of circular derivatives of larger proteins, resulting in improved stability and refolding properties. Here we show that circular proteins can be made reversibly with excellent efficiency by means of a sortase-catalyzed cyclization reaction, requiring only minimal modification of the protein to be circularized.
Sigma-Aldrich
Gly-Gly-Gly, BioUltra, ≥99.0% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
Gly-Gly-Gly
Pricing and availability is not currently available.