Merck
  • Enhanced antioxidant and antityrosinase activities of longan fruit pericarp by ultra-high-pressure-assisted extraction.
Enhanced antioxidant and antityrosinase activities of longan fruit pericarp by ultra-high-pressure-assisted extraction.
Journal of pharmaceutical and biomedical analysis (2009-04-07)
K Nagendra Prasad, Bao Yang, John Shi, Chunyan Yu, Mouming Zhao, Sophia Xue, Yueming Jiang
ABSTRACT
The health benefits of fruits acting against chronic diseases are ascribed to their antioxidant activities which are mainly responsible due to the presence of phenolic compounds. The use of ultra-high-pressure-assisted extraction (UHPE) has shown great advantages for the extraction of these phenolic compounds from longan fruit pericarp (LFP). Studies were carried out to investigate the effects of UHPE at pressures of 200, 300, 400 and 500 MPa on total phenolic contents, extraction yield, antioxidant and antityrosinase activities from LFP. The antioxidant activities of these extracts were analyzed, using various antioxidant models like 1,1-diphenyl-2-picrylhydrazyl (DPPH) radical scavenging activity, total antioxidant capacity and superoxide anion radical scavenging activity. Extract from ultra-high-pressure-assisted extraction at 500MPa (UHPE-500) showed the highest antioxidant activities of all the tested models. In addition, it also showed moderate tyrosinase inhibitory activity. Three phenolic acids, namely gallic acid, ellagic acid, and corilagin were identified and quantified by HPLC. Corilagin content was the highest compared to other phenolic acids identified. UHPE-500 obtained the higher phenolic acid contents compared to other high pressure processing and conventional extractions (CE). Compared with CE, UHPE-500 exhibited good extraction effectiveness in terms of higher extraction yields with high phenolic contents and also with higher antioxidant and antityrosinase activities.
Product Description
Supelco
Corilagin, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Corilagin
Pricing and availability is not currently available.