Merck
Intra-patient reproducibility of myocardial SPECT imaging with 201Tl.
Journal of nuclear cardiology : official publication of the American Society of Nuclear Cardiology (2009-01-20)
Barbra E Backus, Frederik A Verburg, R Leo Romijn, Mark W Konijnenberg, Freek J Beekman, J Fred Verzijlbergen
ABSTRACT
To define the physical and clinical reproducibility of (201)Tl myocardial perfusion SPECT (MPS), this study assesses the variation between two repeated rest (201)Tl MPS with repositioning only, with a two-hour time interval and with phantom measurements as a reference. Three anthropomorphic thorax phantoms were filled with (201)Tl. For each phantom five repeated (201)Tl MPS were obtained. In addition, in 20 patients repeated (201)Tl rest-MPS and in 26 patients early and delayed (201)Tl rest-MPS were performed. Quantitative analysis was done using MunichHeart. Statistical methods were used to calculate variability. Visual analysis was performed by 2 independent observers. The average variation between repeated phantom MPS was 0.5% (95% confidence interval (CI): -0.4% to 1.4%). For patient scans this was -5.0% (95% CI: -2.5% to -7.5%) and between early and delayed (201)Tl MPS -15.5% (95% CI: -11.7% to -19.3%). Visual assessment revealed no clinical significant differences between rest (201)Tl and repeated or delayed (201)Tl MPS. Repositioning in phantom (201)Tl MPS does not cause significant variation. Repeated (201)Tl MPS in patients shows 5.0% decrease of (201)Tl in 30 minutes, which increases to 15% during a two-hour time interval without quantitative or visual regional differences. This decrease indicates a time-related washout of (201)Tl, but does not change clinical diagnosis.
Sigma-Aldrich
Thallium(I) chloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Thallium(I) chloride, 99%
Pricing and availability is not currently available.