Merck
  • Acute central administration of immepip, a histamine H3 receptor agonist, suppresses hypothalamic histamine release and elicits feeding behavior in rats.
Acute central administration of immepip, a histamine H3 receptor agonist, suppresses hypothalamic histamine release and elicits feeding behavior in rats.
Brain research bulletin (2009-01-20)
Seiichi Chiba, Emi Itateyama, Toshiie Sakata, Hironobu Yoshimatsu
ABSTRACT
Histamine suppresses feeding behavior via histamine H1 receptors in the hypothalamus. This study was performed to examine whether the acute reduction of histamine release in the hypothalamus caused by immepip, a histamine H3 agonist, modulates the feeding behavior of rats. Rats had a catheter implanted in the third cerebral ventricle (i3v) and were given central injections of phosphate-buffered-saline or immepip (100-300 pmol/rat). Following the i3v administration of immepip, the rats developed dose-dependent hypokinesia within 10 min of administration. Next to hypokinesia, the rats showed significant dose-dependent feeding behavior. High-performance liquid chromatography (HPLC) confirmed the reduction in histamine release in the hypothalamus of rats following i3v administration of immepip. These results suggest that i3v administration of immepip, an H3 receptor agonist, suppresses hypothalamic histamine release and elicits feeding behavior in rats.
Sigma-Aldrich
1-Methylhistamine dihydrochloride, ≥98% (TLC), powder