- C18, C8, and perfluoro reversed phases on diamond for solid-phase extraction.
C18, C8, and perfluoro reversed phases on diamond for solid-phase extraction.
In spite of advances in solid-phase extraction (SPE) technology there are certain disadvantages to current SPE silica-based, column packings. The pH range over which extraction can occur is limited and each column is generally only used once. New diamond-based reversed SPE phases (C(18), C(8), and perfluorinated) were developed in our laboratories. Studies were done which show that these phases do not have the same limitations as traditional silica-based stationary phases. The synthesis and properties of these diamond-based phases are presented, and the stability, percent recovery, and column capacity are given for the C(18) phase.