Merck
Dynamic nature of the ligustilide complex.
Journal of natural products (2008-09-11)
Andreas Schinkovitz, Samuel M Pro, Matthew Main, Shao-Nong Chen, Birgit U Jaki, David C Lankin, Guido F Pauli
ABSTRACT
Monomeric phthalides such as Z-ligustilide (1) and Z-butylidenephthalide (2) are major constituents of medicinal plants of the Apiaceae family. While 1 has been associated with a variety of observed biological effects, it is also known for its instability and rapid chemical degradation. For the purpose of isolating pure 1 and 2, a gentle and rapid two-step countercurrent isolation procedure was developed. From a supercritical CO2 fluid extract of Angelica sinensis roots, the phthalides were isolated with high GC-MS purities of 99.4% for 1 and 98.9% for 2 and consistently lower qHNMR purities of 98.1% and 96.4%, respectively. Taking advantage of molarity-based qHNMR methodology, a time-resolved study of the dynamic changes and residual complexity of pure 1 was conducted. GC-MS and (qH)NMR analysis of artificially degraded 1 provided evidence for the phthalide degradation pathways and optimized storing conditions. Parallel qHNMR analysis led to the recognition of variations in time- and process-dependent sample purity and has impact on the overall assessment of time-dependent changes in complex natural products systems. The study underscores the importance of independent quantitative monitoring as a prerequisite for the biological evaluation of labile natural products such as monomeric phthalides.