Merck
Fragrance material review on 3,7-dimethyl-1-octanol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
A Lapczynski, S P Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of 3,7-dimethyl-1-octanol when used as a fragrance ingredient is presented.
Sigma-Aldrich
3,7-Dimethyl-1-octanol, ≥98%
Pricing and availability is not currently available.