Merck
Fragrance material review on laevo-carveol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
S P Bhatia, D McGinty, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of laevo-carveol when used as a fragrance ingredient is presented.
Sigma-Aldrich
L-Carveol, mixture of cis and trans, ≥95%, FG
Sigma-Aldrich
(−)-Carveol, mixture of isomers, 97%