Merck
  • Determination of alpha- and beta-amanitin in clinical urine samples by Capillary Zone Electrophoresis.
Determination of alpha- and beta-amanitin in clinical urine samples by Capillary Zone Electrophoresis.
Journal of pharmaceutical and biomedical analysis (2008-05-27)
V A Robinson-Fuentes, J L Jaime-Sánchez, L García-Aguilar, M Gómez-Peralta, M S Vázquez-Garcidueñas, G Vázquez-Marrufo
ABSTRACT
Amanitins are toxins found in species of the mushroom genera Amanita, Lepiota and Galerina. Intoxication after ingestion of these mushrooms can be fatal with an estimated 20% of mortality rate. An early diagnosis is necessary in order to avoid invasive and expensive therapy and to improve patient's prognosis. In this paper, a Capillary Zone Electrophoresis method was developed and validated to determine alpha- and beta-amanitin in urine in less than 7 min using 5 mM, pH 10 borate buffer as background electrolyte. The separation conditions were: capillary: 75 microm I.D., 41 cm effective length, 48 cm total length, 25 degrees C, 20 KV and PDA detection at 214 nm. Sample treatment for analysis only required urine dilution in background electrolyte. The method was validated following established criteria and was found to be selective, linear in the range 5-100 ng/ml. Intra- and inter-day precision and accuracy were within required limits. Limit of detection (LOD) and limit of quantification (LOQ) were 1.5 and 5 ng/ml, respectively. Eight urine samples from suspected cases of intoxication with amanitins were analyzed after 2 years of storage at -20 degrees C, and beta-amanitin was determined in two samples with concentrations of 53 and 65 ng/ml, respectively. The method here described includes the use of non-aggressive reagents to the capillary or the system and is the first Capillary Electrophoresis method used to determine amanitins in clinical samples.
Sigma-Aldrich
β-Amanitin from Amanita phalloides, ≥85% (HPLC)
Pricing and availability is not currently available.