Merck
  • p-dimethylaminocinnamaldehyde derivatization for colorimetric detection and HPLC-UV/vis-MS/MS identification of indoles.
p-dimethylaminocinnamaldehyde derivatization for colorimetric detection and HPLC-UV/vis-MS/MS identification of indoles.
Archives of biochemistry and biophysics (2008-04-22)
Patrick R Porubsky, Emily E Scott, Todd D Williams
ABSTRACT
Cytochrome P450 2A13 (CYP2A13) is a lung specific enzyme known to activate the potent tobacco procarcinogen 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) into two carcinogenic metabolites. CYP2A13 has been crystallized and X-ray diffraction experiments illuminated the structure of this enzyme, but with an unknown ligand present in the enzyme active site. This unknown ligand was suspected to be indole but a selective method had to be developed to differentiate among indole and its metabolites in the protein sample. We successfully modified a microbiological colorimetric assay to spectrophotometrically differentiate between indole and a number of possible indole metabolites in nanomolar concentrations by derivatization with p-dimethylaminocinnamaldehyde (DMACA). Further differentiation of indoles was made by mass spectrometry (HPLC-UV/vis-MS/MS) utilizing the chromophore generated in the DMACA conjugation as a UV signature for HPLC detection. The ligand in the crystallized protein was identified as unsubstituted indole, which facilitated refinement of two alternate conformations in the CYP2A13 crystal structure active site.
Product Description
Millipore
Kovac′s reagent for indoles, suitable for microbiology
Pricing and availability is not currently available.
Millipore
DMACA Reagent, suitable for microbiology
Pricing and availability is not currently available.
Sigma-Aldrich
4-(Dimethylamino)cinnamaldehyde, chromogenic reagent for indoles and flavanols
Pricing and availability is not currently available.