- Flexible graphene films via the filtration of water-soluble noncovalent functionalized graphene sheets.
Flexible graphene films via the filtration of water-soluble noncovalent functionalized graphene sheets.
Journal of the American Chemical Society (2008-04-11)
Yuxi Xu, Hua Bai, Gewu Lu, Chun Li, Gaoquan Shi
PMID18399634
ABSTRACT
Flexible graphene films were prepared by the filtration of water-soluble noncovalently functionalized graphene sheets with pyrenebutyrate. The work presented here will not only open a new way for preparing water-soluble graphene dispersions but also provide a general route for fabricating conducting films based on graphene.