Merck
  • Insulin-secretagogue activity of p-methoxycinnamic acid in rats, perfused rat pancreas and pancreatic beta-cell line.
Insulin-secretagogue activity of p-methoxycinnamic acid in rats, perfused rat pancreas and pancreatic beta-cell line.
Basic & clinical pharmacology & toxicology (2008-03-19)
Sirintorn Yibchok-anun, Sirichai Adisakwattana, Preecha Moonsan, Walter H Hsu
ABSTRACT
This study investigated the effect of p-methoxycinnamic acid (p-MCA) on plasma glucose and insulin concentrations in normal and streptozotocin-induced diabetic rats. In both fasting and glucose-loading conditions, an oral administration of p-MCA (40-100 mg/kg) significantly decreased plasma glucose and also increased plasma insulin concentrations in both normal and diabetic rats. The onset of the p-MCA-induced antihyperglycaemia/hypoglycaemia was observed at 1 hr after administration. In perfused rat pancreas, p-MCA (10-100 microM) stimulated insulin secretion about 1.4- and 3.1-fold of basal-control group. In addition, p-MCA (10 microM) enhanced glucose-induced insulin secretion. Moreover, p-MCA stimulated insulin secretion and increased intracellular Ca(2+) concentration ([Ca(2+)](i)) in insulinoma-1 cells. Taken together, our findings suggested that p-MCA exerted antihyperglycaemic/hypoglycaemic effect by stimulating insulin secretion from pancreas and could be developed into a new potential for therapeutic agent used in type 2 diabetic patients.
Sigma-Aldrich
4-Methoxycinnamic acid, predominantly trans, 99%
Pricing and availability is not currently available.