Merck
  • Inhibitory effects of substrate and product on the carvone biotransformation activity of Rhodococcus erythropolis.
Inhibitory effects of substrate and product on the carvone biotransformation activity of Rhodococcus erythropolis.
Biotechnology letters (2008-03-11)
Jenna L E Morrish, Andrew J Daugulis
ABSTRACT
The aqueous substrate and product toxicity thresholds in the microbial biotransformation of (-)-trans-carveol to the fragrance/flavor compound (R)-(-)-carvone by Rhodococcus erythropolis were determined. Above aqueous phase concentrations of approx. 500 mg carveol/l and 200-600 mg carvone/l, the biotransformation activity of the biocatalyst was inhibited. This biotransformation was undertaken in a single aqueous phase 3 l [corrected] reactor in which a total of 5 ml carveol (mixture of isomers) was added before the biotransformation rate decreased significantly. The carvone volumetric productivity was 31 mg/lh. Although the growth of the organism post-exposure was not affected, dramatic morphological changes in response to the accumulation of the inhibitory substrate and product were observed.
Sigma-Aldrich
L-Carveol, mixture of cis and trans, ≥95%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
(−)-Carveol, mixture of isomers, 97%
Pricing and availability is not currently available.