Merck
  • Scalable synthesis of the VEGF-R2 kinase inhibitor JNJ-17029259 using ultrasound-mediated addition of MeLi-CeCl3 to a nitrile.
Scalable synthesis of the VEGF-R2 kinase inhibitor JNJ-17029259 using ultrasound-mediated addition of MeLi-CeCl3 to a nitrile.
The Journal of organic chemistry (2008-01-04)
Michael Reuman, Sandra Beish, Jeremy Davis, Mark J Batchelor, Martin C Hutchings, David F C Moffat, Peter J Connolly, Ronald K Russell
ABSTRACT
The preparation of the selective VEGF-R2 kinase inhibitor 10 (JNJ-17029259) is described in which the key precursor, 4-(5-isoxazolyl)benzonitrile, undergoes clean transformation to the corresponding cumylamine derivative with CeCl(3)-MeLi in THF. This high-yielding cerium mediated transformation is robust, reproducible, and readily scalable based on a requirement for the anhydrous CeCl(3) to be milled and subjected to ultrasound treatment prior to addition of methyllithium.
Sigma-Aldrich
Cerium(III) chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Cerium(III) chloride, AnhydroBeads, −10 mesh, 99.9%
Sigma-Aldrich
Cerium(III) chloride heptahydrate, 99.9% trace metals basis
Sigma-Aldrich
Cerium(III) chloride heptahydrate, purum p.a., ≥98.0% (AT)
Sigma-Aldrich
Cerium(III) chloride heptahydrate, 99.999% trace metals basis