Merck
  • Magnetic nanoparticle-supported proline as a recyclable and recoverable ligand for the CuI catalyzed arylation of nitrogen nucleophiles.
Magnetic nanoparticle-supported proline as a recyclable and recoverable ligand for the CuI catalyzed arylation of nitrogen nucleophiles.
Chemical communications (Cambridge, England) (2007-11-16)
Gagan Chouhan, Dashan Wang, Howard Alper
ABSTRACT
Magnetic nanoparticle-supported proline ligand was prepared and used for the CuI catalyzed Ullmann-type coupling reactions of aryl/heteroaryl bromides with various nitrogen heterocycles to form the corresponding N-aryl products in good to excellent yields; furthermore, this magnetic nanoparticle-supported proline ligand could be readily separated using an external magnet and reused without significant loss of activity.
Sigma-Aldrich
4′-Bromoacetophenone, 98%
Pricing and availability is not currently available.