- Eriochrome black T as a dye for agarose gel electrophoresis.
Eriochrome black T as a dye for agarose gel electrophoresis.
Acta biochimica Polonica (1991-01-01)
M Obuchowski, G Wegrzyn
PMID1796700
ABSTRACT
We found that it is possible to use eriochrome black T as a dye for agarose gel electrophoresis of DNA. The presence of the dye does not change the migration rate of DNA and does not influence the electrophoretical picture.