Merck
Myxothiazol, a new antibiotic interfering with respiration.
Antimicrobial agents and chemotherapy (1981-04-01)
G Thierbach, H Reichenbach
ABSTRACT
Myxothiazol, a new antibiotic from the myxobacterium Myxococcus fulvus, inhibited the growth of many yeasts and fungi at concentrations between 0.01 and 3 micrograms/ml. It was generally inactive against bacteria. The inhibitory effect was cytostatic. With Candida albicans, Saccharomyces cerevisiae, and Mucor hiemalis, the growth inhibition was neutralized by glucose. Soon after being added to a cell suspension, the compound almost completely blocked oxygen consumption.
Sigma-Aldrich
Myxothiazol, from Myxococcus fulvus Mx f85, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)