Merck
[A model study of pesticide biodegradation in soil].
Izvestiia Akademii nauk. Seriia biologicheskaia (2007-03-14)
J Bieganska
ABSTRACT
This study addresses the efficiency of microbial preparations to degrade pesticide residues in soil. A method to degrade pesticides DNOC and pendimethalin using Pseudomonas and Arthrobacter bacteria with a fertilizer is described.
Product Description
Supelco
DNOC, PESTANAL®, analytical standard