Merck
  • Conformation and structure of bis(glycyl-L-aspartic acid) oxalate 0.4-hydrate.
Conformation and structure of bis(glycyl-L-aspartic acid) oxalate 0.4-hydrate.
Acta crystallographica. Section C, Crystal structure communications (2007-01-09)
R Chitra, Vijay Thiruvenkatam, R R Choudhury, M V Hosur, T N Guru Row
ABSTRACT
The title bis(glycyl-L-aspartic acid) oxalate complex {systematic name: bis[2-(2-ammonioacetamido)butanedioic acid] oxalate 0.4-hydrate}, 2C6H11N2O5+.C2O4(2-).4H2O, crystallizes in a triclinic space group with the planar peptide unit in a trans conformation. The asymmetric unit consists of two glycyl-L-aspartic acid molecules with positively charged amino groups and neutral carboxyl groups, and an oxalate dianion. The twist around the C-Calpha bond indicates that both the peptide molecules adopt extended conformations, while the twist around the N-Calpha bond shows that one has a folded and the other a semi-extended state. The present complex can be described as an inclusion compound with the dipeptide molecule as the host and the oxalate anion as the guest. The usual head-to-tail sequence of aggregation is not observed in this complex, as is also the case with the glycyl-L-aspartic acid dihydrate molecule. The study of aggregation and interaction patterns in binary systems is the first step towards understanding more complex phenomena. This further leads to results that are of general interest in bimolecular aggregation.
Sigma-Aldrich
Gly-Asp, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Gly-DL-Asp
Pricing and availability is not currently available.